Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Morpholinonicotinic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

043986-500MG 043986-1G 043986-5G
101863-530EA 92.31 USD InStock
101863-530 101863-532 101863-534
6-Morpholinonicotinic acid ≥95%
6-Morpholinonicotinic acid
Hazard: Irritant


Formula: C₁₀H₁₂N₂O₃
Melting Pt: 258-261°C
MDL Number: MFCD02682034
CAS Number: 120800-52-4
Order Now
Specification Test Results
Melting point 258-261°C

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty