Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyano-3-(4-methylphenyl)acrylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

044009-500MG 044009-1G 044009-5G
101863-654EA 181.54 USD InStock
101863-654 101863-656 101863-658
2-Cyano-3-(4-methylphenyl)acrylic acid ≥95%
2-Cyano-3-(4-methylphenyl)acrylic acid
Hazard: Irritant


Formula: C₁₁H₉NO₂
Melting Pt: 199…202 °C
MDL Number: MFCD01313927
CAS Number: 20374-48-5
Order Now
Specification Test Results
Melting point 199-202°C

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty