Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Ethoxycarbonyl)-4,4-diphenyl-3-butenoic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

044048-500MG 044048-1G 044048-5G
101863-856EA 181.54 USD InStock
101863-856 101863-858 101863-860
3-(Ethoxycarbonyl)-4,4-diphenyl-3-butenoic acid ≥95%
3-(Ethoxycarbonyl)-4,4-diphenyl-3-butenoic acid
Hazard: Irritant


Formula: C₁₉H₁₈O₄
Melting Pt: 128…130 °C
MDL Number: MFCD00215361
CAS Number: 5438-22-2
Order Now
Specification Test Results
Melting point 128-130°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty