Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-(4-nitrophenyl)acetonitrile ≥95%

Supplier: MATRIX SCIENTIFIC MS

044079-500MG 044079-1G 044079-5G
101864-020EA 375.38 USD InStock
101864-020 101864-022 101864-024
2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-(4-nitrophenyl)acetonitrile ≥95%
2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-(4-nitrophenyl)acetonitrile
Hazard: Irritant


Formula: C₁₄H₇ClF₃N₃O₂
Melting Pt: 110…112 °C
MDL Number: MFCD00172215
CAS Number: 213994-29-7
Order Now
Specification Test Results
Melting point 110-112°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty