Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenyl-5-(1H-pyrazol-3-yl)-1,3-thiazole ≥95%

Supplier: MATRIX SCIENTIFIC MS

044149-500MG 044149-1G 044149-5G
101864-410EA 556.92 USD InStock
101864-410 101864-412 101864-414
2-Phenyl-5-(1H-pyrazol-3-yl)-1,3-thiazole ≥95%
2-Phenyl-5-(1H-pyrazol-3-yl)-1,3-thiazole
Hazard: Irritant


Formula: C₁₂H₉N₃S
Melting Pt: 240…243 °C
MDL Number: MFCD00974300
CAS Number: 318234-44-5
Order Now
Specification Test Results
Melting point 240-243°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty