Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-{3-[(2,4-Dichlorobenzyl)oxy]phenyl}-1-ethanone ≥95%

Supplier: MATRIX SCIENTIFIC MS

044203-500MG 044203-1G 044203-5G
101864-698EA 181.54 USD InStock
101864-698 101864-700 101864-702
1-{3-[(2,4-Dichlorobenzyl)oxy]phenyl}-1-ethanone ≥95%
1-{3-[(2,4-Dichlorobenzyl)oxy]phenyl}-1-ethanone
Hazard: Irritant


Formula: C₁₅H₁₂Cl₂O₂
MW: 295.17 g/mol
Melting Pt: 71…73 °C
MDL Number: MFCD03001258
CAS Number: 400878-28-6
Order Now
Specification Test Results
Melting point 71-73°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty