Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Iodoimidazo[1,2-a]pyridine-2-carboxylic acid 95

Supplier: MATRIX SCIENTIFIC MS

044208-500MG 044208-1G
101864-728EA 375.38 USD InStock
101864-728 101864-730
6-Iodoimidazo[1,2-a]pyridine-2-carboxylic acid 95
6-Iodoimidazo[1,2-a]pyridine-2-carboxylic acid
Hazard: Irritant


Formula: C₈H₅IN₂O₂
MW: 288.05 g/mol
Melting Pt: 244…246 °C
MDL Number: MFCD02186210
CAS Number: 478040-59-4
Order Now
Specification Test Results
Melting point 244-246°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty