Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methyl-2-(methylsulfanyl)-5-pyrimidinecarboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

044220-500MG 044220-1G 044220-5G
101864-788EA 556.92 USD InStock
101864-788 101864-790 101864-792
4-Methyl-2-(methylsulfanyl)-5-pyrimidinecarboxylic acid ≥95%
4-Methyl-2-(methylsulfanyl)-5-pyrimidinecarboxylic acid
Hazard: Irritant


Formula: C₇H₈N₂O₂S
Melting Pt: 169…171 °C
MDL Number: MFCD02187391
CAS Number: 98276-75-6
Order Now
Specification Test Results
Melting point 169-171°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty