Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(tert-Butyl)cyclohexanone oxime ≥95%

Supplier: MATRIX SCIENTIFIC MS

044271-500MG 044271-1G 044271-5G
101865-026EA 181.54 USD InStock
101865-026 101865-028 101865-030
4-(tert-Butyl)cyclohexanone oxime ≥95%
4-(tert-Butyl)cyclohexanone oxime
Hazard: Irritant


Formula: C₁₀H₁₉NO
Melting Pt: 137…139 °C
CAS Number: 4701-98-8
Order Now
Specification Test Results
Melting point 137-139°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty