Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Oxo-3-[3-(trifluoromethyl)phenyl]3,4-dihydro-1-phthalazinecarboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

045544-500MG 045544-1G 045544-5G
101871-394EA 375.38 USD InStock
101871-394 101871-396 101871-398
4-Oxo-3-[3-(trifluoromethyl)phenyl]3,4-dihydro-1-phthalazinecarboxylic acid ≥95%
4-Oxo-3-[3-(trifluoromethyl)phenyl]3,4-dihydro-1-phthalazinecarboxylic acid
Hazard: Irritant


Formula: C₁₆H₉F₃N₂O₃
Melting Pt: 210…213 °C
MDL Number: MFCD00138613
CAS Number: 339021-26-0
Order Now
Specification Test Results
Melting point 210-213°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty