Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzyl-5-chloro-6-oxo-1,6-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

045561-500MG 045561-1G 045561-5G
101871-482EA 556.92 USD InStock
101871-482 101871-484 101871-486
1-Benzyl-5-chloro-6-oxo-1,6-dihydro-3-pyridine carboxylic acid ≥95%
1-Benzyl-5-chloro-6-oxo-1,6-dihydro-3-pyridine carboxylic acid
Hazard: Irritant


Formula: C₁₃H₁₀ClNO₃
MW: 263.68 g/mol
Melting Pt: 208…209 °C
MDL Number: MFCD00232709
CAS Number: 338982-47-1
Order Now
Specification Test Results
Melting point 208-209°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty