Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[2-(4-Methoxyphenyl)-1,3-thiazol-4-yl]acetic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

045606-500MG 045606-1G 045606-5G
101871-712EA 38.46 USD InStock
101871-712 101871-714 101871-716
2-[2-(4-Methoxyphenyl)-1,3-thiazol-4-yl]acetic acid ≥95%
2-[2-(4-Methoxyphenyl)-1,3-thiazol-4-yl]acetic acid
Hazard: Irritant


Formula: C₁₂H₁₁NO₃S
MW: 249.29 g/mol
Melting Pt: 105…107 °C
MDL Number: MFCD01316979
CAS Number: 23353-14-2
Order Now
Specification Test Results
Melting point 105-107°C

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty