Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-[3-(Trifluoromethyl)benzyl]urea ≥95%

Supplier: MATRIX SCIENTIFIC MS

045621-500MG 045621-1G 045621-5G
101871-780EA 181.54 USD InStock
101871-780 101871-782 101871-784
N-[3-(Trifluoromethyl)benzyl]urea ≥95%
N-[3-(Trifluoromethyl)benzyl]urea
Hazard: Irritant


Formula: C₉H₉F₃N₂O
MW: 218.18 g/mol
Melting Pt: 118…120 °C
MDL Number: MFCD01314643
CAS Number: 296277-59-3
Order Now
Specification Test Results
Melting point 118-120°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty