Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-[(aminocarbonyl)amino]acetate ≥95%

Supplier: MATRIX SCIENTIFIC MS

045622-500MG 045622-1G 045622-5G
101871-786EA 307.69 USD InStock
101871-786 101871-788 101871-790
Ethyl-2-[(aminocarbonyl)amino]acetate ≥95%
Ethyl-2-[(aminocarbonyl)amino]acetate
Hazard: Irritant


Formula: C₅H₁₀N₂O₃
Melting Pt: 146…148 °C
MDL Number: MFCD00025456
CAS Number: 6293-20-5
Order Now
Specification Test Results
Melting point 146-148°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty