Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Morpholino)-3-nitropyridine ≥95%

Supplier: MATRIX SCIENTIFIC MS

045672-500MG 045672-1G 045672-5G
101872-038EA 278.46 USD InStock
101872-038 101872-040 101872-042
2-(4-Morpholino)-3-nitropyridine ≥95%
2-(4-Morpholino)-3-nitropyridine
Hazard: Irritant


Formula: C₉H₁₁N₃O₃
MW: 209.2019 g/mol
Melting Pt: 91…94 °C
MDL Number: MFCD00452814
CAS Number: 24255-27-4
Order Now
Specification Test Results
Melting point 91-94°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty