Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-3-[4-(methylsulfanyl)phenyl]propanoic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

045682-500MG 045682-1G 045682-5G
101872-084EA 278.46 USD InStock
101872-084 101872-086 101872-088
3-Amino-3-[4-(methylsulfanyl)phenyl]propanoic acid ≥95%
3-Amino-3-[4-(methylsulfanyl)phenyl]propanoic acid
Hazard: Irritant


Formula: C₁₀H₁₃NO₂S
MW: 211.28 g/mol
Melting Pt: 210…212 °C
MDL Number: MFCD02653333
CAS Number: 412925-13-4
Order Now
Specification Test Results
Melting point 210-212°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty