Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-(tert-butyl)-1-(3-nitro-2-pyridinyl)-1H-pyrazole-5-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

045718-500MG 045718-1G 045718-5G
101872-268EA 375.38 USD InStock
101872-268 101872-270 101872-272
Ethyl-3-(tert-butyl)-1-(3-nitro-2-pyridinyl)-1H-pyrazole-5-carboxylate ≥95%
Ethyl-3-(tert-butyl)-1-(3-nitro-2-pyridinyl)-1H-pyrazole-5-carboxylate
Hazard: Irritant


Formula: C₁₅H₁₈N₄O₄
Melting Pt: 99…101 °C
MDL Number: MFCD05021846
CAS Number: 959572-95-3
Order Now
Specification Test Results
Melting point 99-101°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty