Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-{[5-(Ethoxycarbonyl)-1-methyl-1H-pyrrol-3-yl]carbonyl}benzenecarboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

045746-500MG 045746-1G
101872-406EA 747.69 USD InStock
101872-406 101872-408
2-{[5-(Ethoxycarbonyl)-1-methyl-1H-pyrrol-3-yl]carbonyl}benzenecarboxylic acid ≥95%
2-{[5-(Ethoxycarbonyl)-1-methyl-1H-pyrrol-3-yl]carbonyl}benzenecarboxylic acid
Hazard: Irritant


Formula: C₁₆H₁₅NO₅
Melting Pt: 183…186 °C
MDL Number: MFCD06200987
CAS Number: 120562-90-5
Order Now
Specification Test Results
Melting point 183-186°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty