Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-{4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazino}acetate ≥95%

Supplier: MATRIX SCIENTIFIC MS

045826-500MG 045826-1G 045826-5G
101872-808EA 747.69 USD InStock
101872-808 101872-810 101872-812
Ethyl-2-{4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazino}acetate ≥95%
Ethyl-2-{4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazino}acetate
Hazard: Irritant


Formula: C₁₄H₁₇ClF₃N₃O₂
Melting Pt: 70…72 °C
MDL Number: MFCD00141695
CAS Number: 338979-08-1
Order Now
Specification Test Results
Melting point 70-72°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty