Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidin-2-amine ≥95%

Supplier: MATRIX SCIENTIFIC MS

045828-500MG 045828-1G 045828-5G
101872-820EA 1109.23 USD InStock
101872-820 101872-822 101872-824
6-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidin-2-amine ≥95%
6-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidin-2-amine
Hazard: Irritant


Formula: C₁₃H₁₁ClF₃N₅
Melting Pt: 200…202 °C
MDL Number: MFCD00202227
CAS Number: 338979-12-7
Order Now
Specification Test Results
Melting point 200-202°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty