Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N'-Bis[(dimethylamino)methylene]thiourea ≥95%

Supplier: MATRIX SCIENTIFIC MS

045831-500MG 045831-1G 045831-5G
101872-836EA 181.54 USD InStock
101872-836 101872-838 101872-840
N,N'-Bis[(dimethylamino)methylene]thiourea ≥95%
N,N'-Bis[(dimethylamino)methylene]thiourea
Hazard: Irritant


Formula: C₇H₁₄N₄S
Melting Pt: 187…189 °C
MDL Number: MFCD00173502
CAS Number: 121876-98-0
Order Now
Specification Test Results
Melting point 187-189°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty