Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-(2-bromoacetyl)thiophene ≥95%

Supplier: MATRIX SCIENTIFIC MS

045856-500MG 045856-1G 045856-5G
101872-950EA 556.92 USD InStock
101872-950 101872-952 101872-954
3-Chloro-2-(2-bromoacetyl)thiophene ≥95%
3-Chloro-2-(2-bromoacetyl)thiophene
Hazard: Irritant


Formula: C₆H₄BrClOS
Melting Pt: 59-61°C
MDL Number: MFCD00793892
CAS Number: 256486-71-2
Order Now
Specification Test Results
Melting point 59-61°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty