Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2,2-dimethyl-N-(1,3-thiazol-2-yl)propanamide ≥95%

Supplier: MATRIX SCIENTIFIC MS

045867-500MG 045867-1G 045867-5G
101873-014EA 181.54 USD InStock
101873-014 101873-016 101873-018
3-Chloro-2,2-dimethyl-N-(1,3-thiazol-2-yl)propanamide ≥95%
3-Chloro-2,2-dimethyl-N-(1,3-thiazol-2-yl)propanamide
Hazard: Irritant


Formula: C₈H₁₁ClN₂OS
Melting Pt: 74…76 °C
MDL Number: MFCD00793441
CAS Number: 454473-70-2
Order Now
Specification Test Results
Melting point 74-76°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty