Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]piperazino}aniline hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

045875-500MG 045875-1G 045875-5G
101873-056EA 946.15 USD InStock
101873-056 101873-058 101873-060
4-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]piperazino}aniline hydrochloride ≥95%
4-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]piperazino}aniline hydrochloride
Hazard: Irritant


Formula: C₁₆H₁₇Cl₂F₃N₄
Melting Pt: 230 °C
MDL Number: MFCD00793115
CAS Number: 321848-37-7
Order Now
Specification Test Results
Melting point 230°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty