Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-(4-benzylpiperazino)nicotinate ≥95%

Supplier: MATRIX SCIENTIFIC MS

045905-500MG 045905-1G 045905-5G
101873-188EA 181.54 USD InStock
101873-188 101873-190 101873-192
Methyl-6-(4-benzylpiperazino)nicotinate ≥95%
Methyl-6-(4-benzylpiperazino)nicotinate
Hazard: Irritant


Formula: C₁₈H₂₁N₃O₂
MW: 311.39 g/mol
Melting Pt: 101…104 °C
MDL Number: MFCD01936461
CAS Number: 132521-74-5
Order Now
Specification Test Results
Melting point 101-104°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty