Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2,5-Dimethyl-1H-pyrrol-1-yl)benzenecarbaldehyde ≥95%

Supplier: MATRIX SCIENTIFIC MS

045981-500MG 045981-1G 045981-5G
101873-578EA 278.46 USD InStock
101873-578 101873-580 101873-582
4-(2,5-Dimethyl-1H-pyrrol-1-yl)benzenecarbaldehyde ≥95%
4-(2,5-Dimethyl-1H-pyrrol-1-yl)benzenecarbaldehyde
Hazard: Irritant


Formula: C₁₃H₁₃NO
Melting Pt: 82…85 °C
MDL Number: MFCD00177098
CAS Number: 95337-70-5
Order Now
Specification Test Results
Melting point 82-85°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty