Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-hydroxy-2-thiophenecarboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

045990-1G 045990-5G
101873-626EA 56.92 USD InStock
101873-626 101873-628
Methyl-3-hydroxy-2-thiophenecarboxylate ≥95%
Methyl-3-hydroxy-2-thiophenecarboxylate
Hazard: Irritant


Formula: C₆H₆O₃S
MW: 158.18 g/mol
Boiling Pt: 108 °C (13 Torr)
Melting Pt: 40 °C
MDL Number: MFCD00055642
CAS Number: 5118-06-9
Order Now
Specification Test Results
Melting point 48-51°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty