Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-(5-aminopyridin-2-yl)piperazine-1-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

046032-500MG 046032-1G 046032-5G
101873-822EA 747.69 USD InStock
101873-822 101873-824 101873-826
tert-Butyl-4-(5-aminopyridin-2-yl)piperazine-1-carboxylate ≥95%
tert-Butyl-4-(5-aminopyridin-2-yl)piperazine-1-carboxylate
Hazard: Irritant


Formula: C₁₄H₂₂N₄O₂
Melting Pt: 98…101 °C
MDL Number: MFCD05864841
CAS Number: 119285-07-3
Order Now
Specification Test Results
Melting point 98-101°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty