046054-500MG
046054-1G
046054-5G
101873-936EA
18.46
USD
InStock
101873-936
101873-938
101873-940
5-Amino-3-(4-methoxyphenyl)-1H-pyrazole ≥95%
5-Amino-3-(4-methoxyphenyl)-1H-pyrazole
Purity: 95+%
CAS: 19541-95-8
MDL Number: MFCD00462192
Molecular Formula: C10H11N3O
Molecular Weight: 189,22
Hazard: Irritant