Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Diphenyl-1,3-thiazol-4-ol ≥95%

Supplier: MATRIX SCIENTIFIC MS

046504-500MG 046504-1G 046504-5G
101875-774EA 181.54 USD InStock
101875-774 101875-776 101875-778
2,5-Diphenyl-1,3-thiazol-4-ol ≥95%
2,5-Diphenyl-1,3-thiazol-4-ol
Hazard: Irritant


Formula: C₁₅H₁₁NOS
MW: 253.33 g/mol
Melting Pt: 210…212 °C
MDL Number: MFCD00895725
CAS Number: 59484-42-3
Order Now
Specification Test Results
Melting point 210-212°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty