Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-piperidinobenzenecarbohydrazide ≥95%

Supplier: MATRIX SCIENTIFIC MS

046565-500MG 046565-1G
101876-066EA 375.38 USD InStock
101876-066 101876-068
2-Chloro-4-piperidinobenzenecarbohydrazide ≥95%
2-Chloro-4-piperidinobenzenecarbohydrazide
Hazard: Irritant


Formula: C₁₂H₁₆ClN₃O
Melting Pt: 136…138 °C
MDL Number: MFCD09607913
CAS Number: 952183-28-7
Order Now
Specification Test Results
Melting point 136-138°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty