Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-{2-[3-Fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

047734-500MG 047734-1G 047734-5G
101881-162EA 556.92 USD InStock
101881-162 101881-164 101881-166
2-{2-[3-Fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid ≥95%
2-{2-[3-Fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid
Hazard: Irritant


Formula: C₁₂H₇F₄NO₂S
Melting Pt: 116…118 °C
MDL Number: MFCD09027135
CAS Number: 937602-42-1
Order Now
Specification Test Results
Melting point 116-118°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty