Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[2-Amino-7,8-dihydropyrido[4,3-d]pyrimidin-6(5H)-yl](phenyl)methanone ≥95%

Supplier: MATRIX SCIENTIFIC MS

047777-500MG 047777-1G 047777-5G
101881-386EA 375.38 USD InStock
101881-386 101881-388 101881-390
[2-Amino-7,8-dihydropyrido[4,3-d]pyrimidin-6(5H)-yl](phenyl)methanone ≥95%
[2-Amino-7,8-dihydropyrido[4,3-d]pyrimidin-6(5H)-yl](phenyl)methanone
Hazard: Irritant


Formula: C₁₄H₁₄N₄O
MW: 254.29 g/mol
Melting Pt: 205…207 °C
MDL Number: MFCD09027163
CAS Number: 937604-00-7
Order Now
Specification Test Results
Melting point 205-207°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty