Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Iodo-2-oxo-1-[4-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

047913-500MG 047913-1G 047913-5G
101881-974EA 556.92 USD InStock
101881-974 101881-976 101881-978
5-Iodo-2-oxo-1-[4-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid ≥95%
5-Iodo-2-oxo-1-[4-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid
Hazard: Irritant


Formula: C₁₄H₉F₃INO₃
Melting Pt: 202…205 °C
MDL Number: MFCD09027178
CAS Number: 937604-36-9
Order Now
Specification Test Results
Melting point 202-205°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty