Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Methylsulfonyl)-4-piperidinecarbonyl chloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

047941-500MG 047941-1G
101882-088EA 307.69 USD InStock
101882-088 101882-090
1-(Methylsulfonyl)-4-piperidinecarbonyl chloride ≥95%
1-(Methylsulphonyl)-4-piperidinecarbonyl chloride
Hazard: Irritant/Refrigerate


Formula: C₇H₁₂ClNO₃S
Melting Pt: 81…83 °C
MDL Number: MFCD09865018
CAS Number: 241138-40-9
Order Now
Specification Test Results
Melting point 81-83°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty