Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-methyl-6-(4-methylpiperazino)pyrimidine ≥95%

Supplier: MATRIX SCIENTIFIC MS

048048-500MG 048048-1G 048048-5G
101882-554EA 181.54 USD InStock
101882-554 101882-556 101882-558
4-Chloro-2-methyl-6-(4-methylpiperazino)pyrimidine ≥95%
4-Chloro-2-methyl-6-(4-methylpiperazino)pyrimidine
Hazard: Irritant


Formula: C₁₀H₁₅ClN₄
Melting Pt: 80…82 °C
MDL Number: MFCD09972245
CAS Number: 1017782-75-0
Order Now
Specification Test Results
Melting point 80-82°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty