Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-(6-chloro-2-methyl-4-pyrimidinyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

048049-500MG 048049-1G 048049-5G
101882-560EA 264.62 USD InStock
101882-560 101882-562 101882-564
tert-Butyl-4-(6-chloro-2-methyl-4-pyrimidinyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-(6-chloro-2-methyl-4-pyrimidinyl)tetrahydro-1(2H)-pyrazinecarboxylate
Hazard: Irritant


Formula: C₁₄H₂₁ClN₄O₂
MW: 312.8 g/mol
Melting Pt: 117…119 °C
MDL Number: MFCD09972246
CAS Number: 203519-37-3
Order Now
Specification Test Results
Melting point 117-119°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty