Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Trichlorophenyl-5-bromo-2-thiophenesulfonate ≥95%

Supplier: MATRIX SCIENTIFIC MS

048130-500MG 048130-1G 048130-5G
101882-950EA 278.46 USD InStock
101882-950 101882-952 101882-954
2,4,6-Trichlorophenyl-5-bromo-2-thiophenesulfonate ≥95%
2,4,6-Trichlorophenyl-5-bromo-2-thiophenesulfonate
Hazard: Irritant


Formula: C₁₀H₄BrCl₃O₃S₂
Melting Pt: 54…56 °C
MDL Number: MFCD12498726
CAS Number: 1171919-31-5
Order Now
Specification Test Results
Melting point 54-56°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty