Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Trichlorophenyl-1-ethanesulfonate ≥95%

Supplier: MATRIX SCIENTIFIC MS

048132-500MG 048132-1G 048132-5G
101882-962EA 278.46 USD InStock
101882-962 101882-964 101882-966
2,4,6-Trichlorophenyl-1-ethanesulfonate ≥95%
2,4,6-Trichlorophenyl 1-ethanesulphonate
Hazard: Irritant


Formula: C₈H₇Cl₃O₃S
Melting Pt: 72-74°C
MDL Number: MFCD12498727
CAS Number: 934986-60-4
Order Now
Specification Test Results
Melting point 72-74°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty