Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Propan-2-yl N-[(2S)-7-bromo-1H,2H,3H,4H-cyclopenta[b]indol-2-yl]carbamate ≥95%

Supplier: MATRIX SCIENTIFIC MS

048539-500MG 048539-1G 048539-5G
101884-284EA 921.54 USD InStock
101884-284 101884-286 101884-288
Propan-2-yl N-[(2S)-7-bromo-1H,2H,3H,4H-cyclopenta[b]indol-2-yl]carbamate ≥95%
Propan-2-yl N-[(2S)-7-bromo-1H,2H,3H,4H-cyclopenta[b]indol-2-yl]carbamate
Hazard: Irritant


Formula: C₁₅H₁₇BrN₂O₂
MW: 337.22 g/mol
Melting Pt: 174…176 °C
MDL Number: MFCD13193067
CAS Number: 1212283-31-2
Order Now
Specification Test Results
Melting point 174-176°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty