Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-3-methyl-1H-pyrazole-4-carbonitrile ≥97%

Supplier: MATRIX SCIENTIFIC MS

048568-500MG 048568-1G 048568-5G
101884-412EA 115.38 USD InStock
101884-412 101884-414 101884-416
5-Amino-3-methyl-1H-pyrazole-4-carbonitrile ≥97%
5-Amino-3-methyl-1H-pyrazole-4-carbonitrile
Hazard: Irritant


Formula: C₅H₆N₄
MW: 122.13 g/mol
Melting Pt: 161…163 °C
MDL Number: MFCD02020550
CAS Number: 5453-07-6
Order Now
Specification Test Results
Melting point 161-163°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty