Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-2-(methylamino)-3-nitrobenzenecarbonitrile ≥95%

Supplier: MATRIX SCIENTIFIC MS

048591-500MG 048591-1G
101884-502EA 375.38 USD InStock
101884-502 101884-504
6-Chloro-2-(methylamino)-3-nitrobenzenecarbonitrile ≥95%
6-Chloro-2-(methylamino)-3-nitrobenzenecarbonitrile
Hazard: Irritant


Formula: C₈H₆ClN₃O₂
Melting Pt: 165…168 °C
MDL Number: MFCD09835343
CAS Number: 333458-39-2
Order Now
Specification Test Results
Melting point 165-168°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty