Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methoxy-6-(trifluoromethyl)-1,3-dihydro-2H-indol-2-one ≥95%

Supplier: MATRIX SCIENTIFIC MS

048616-500MG 048616-1G 048616-5G
101884-598EA 1109.23 USD InStock
101884-598 101884-600 101884-602
5-Methoxy-6-(trifluoromethyl)-1,3-dihydro-2H-indol-2-one ≥95%
5-Methoxy-6-(trifluoromethyl)-1,3-dihydro-2H-indol-2-one
Hazard: Irritant


Formula: C₁₀H₈F₃NO₂
MW: 231.18 g/mol
Melting Pt: 226…228 °C
MDL Number: MFCD12922678
CAS Number: 1190198-26-5
Order Now
Specification Test Results
Melting point 226-228°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty