Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Benzyloxy)-5-nitrobenzenecarbohydrazide ≥95%

Supplier: MATRIX SCIENTIFIC MS

048653-500MG 048653-1G
101884-742EA 375.38 USD InStock
101884-742 101884-744
2-(Benzyloxy)-5-nitrobenzenecarbohydrazide ≥95%
2-(Benzyloxy)-5-nitrobenzenecarbohydrazide
Hazard: Irritant


Formula: C₁₄H₁₃N₃O₄
Melting Pt: 126…128 °C
MDL Number: MFCD13193077
CAS Number: 1192263-76-5
Order Now
Specification Test Results
Melting point 126-128°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty