Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-hydroxybenzenecarbonitrile ≥95%

Supplier: MATRIX SCIENTIFIC MS

048695-1G 048695-5G
101884-904EA 75.38 USD InStock
101884-904 101884-906
2-Fluoro-5-hydroxybenzenecarbonitrile ≥95%
2-Fluoro-5-hydroxybenzenecarbonitrile
Hazard: Irritant


Formula: C₇H₄FNO
MW: 137.1100 g/mol
Melting Pt: 111…113 °C
MDL Number: MFCD11846514
CAS Number: 104798-53-0
Order Now
Specification Test Results
Melting point 111-113°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty