Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-chloro-4-hydrazinonicotinate ≥95%

Supplier: MATRIX SCIENTIFIC MS

048702-500MG 048702-1G
101884-934EA 747.69 USD InStock
101884-934 101884-936
Methyl-6-chloro-4-hydrazinonicotinate ≥95%
Methyl-6-chloro-4-hydrazinonicotinate
Hazard: Irritant


Formula: C₇H₈ClN₃O₂
Melting Pt: 150…152 °C
MDL Number: MFCD13193108
CAS Number: 65973-40-2
Order Now
Specification Test Results
Melting point 150-152°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty