Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-3-methoxypyrrolo[1,2-a]quinoxaline ≥95%

Supplier: MATRIX SCIENTIFIC MS

056378-500MG 056378-1G
101910-264EA 538.46 USD InStock
101910-264 101910-266
6-Chloro-3-methoxypyrrolo[1,2-a]quinoxaline ≥95%
6-Chloro-3-methoxypyrrolo[1,2-a]quinoxaline
Hazard: Irritant


Formula: C₁₂H₉ClN₂O
Melting Pt: 134…135 °C
MDL Number: MFCD28050016
CAS Number: 160657-08-9
Order Now
Specification Test Results
Melting point 134-135°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty