Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-iodo-6-methoxypyridine-3-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

056432-500MG 056432-1G
101910-480EA 387.69 USD InStock
101910-480 101910-482
Methyl-5-iodo-6-methoxypyridine-3-carboxylate ≥95%
Methyl-5-iodo-6-methoxypyridine-3-carboxylate
Hazard: Irritant


Formula: C₈H₈INO₃
Melting Pt: 113…115 °C
MDL Number: MFCD16710294
CAS Number: 1242268-18-3
Order Now
Specification Test Results
Melting point 113-115°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty