Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzoyl-4-phenylsemicarbazide

Supplier: MATRIX SCIENTIFIC MS

056892-5G 056892-25G
101911-616EA 116.92 USD InStock
101911-616 101911-618
1-Benzoyl-4-phenylsemicarbazide
1-Benzoyl-4-phenylsemicarbazide
Hazard: Irritant


Formula: C₁₄H₁₃N₃O₂
Melting Pt: 220…222 °C
MDL Number: MFCD00088033
CAS Number: 1152-32-5
Order Now
Specification Test Results
Melting point 220-222°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty