Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl phthalimidomalonate ≥98%

Supplier: MATRIX SCIENTIFIC MS

057017-5G 057017-25G
101912-056EA 170.77 USD InStock
101912-056 101912-058
Diethyl phthalimidomalonate ≥98%
Diethyl phthalimidomalonate
Hazard: Irritant


Formula: C₁₅H₁₅NO₆
MW: 305.29 g/mol
Melting Pt: 75…77 °C
MDL Number: MFCD00065025
CAS Number: 5680-61-5
Order Now
Specification Test Results
Melting point 75-77°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty